sec-Butyl acetate ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1-Methylpropyl acetate
A0025-500ML
A0025-25ML
TCA0025-500MLEA
129
USD
TCA0025-500ML
TCA0025-25ML
sec-Butyl acetate ≥98.0%
sec-Butyl acetate
CAS Number: 105-46-4
MDL Number: MFCD00009328
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 18
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Acetic Acid sec-Butyl Ester
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 112 °C Melting Pt: -98.9 °C Density: 0.87 (20 °C) Flash Pt: 18 °C |
MDL Number:
MFCD00009328 CAS Number: 105-46-4 EINECS: 203-300-1 UN: 1123 ADR: 3,II Merck Index: 13,01535 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...