Benzyl acetate ≥99.0%

Supplier: TCI America

A0022-25G A0022-500G
TCA0022-025GEA 21.26 USD
TCA0022-025G TCA0022-500G
Benzyl acetate ≥99.0%
Benzyl acetate

CAS Number: 140-11-4
MDL Number: MFCD00008712
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Melting point (°C): -51
Flash Point (°C): 102
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Acetic Acid Benzyl Ester

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 214 °C
Melting Pt: -51 °C
Density: 1.06 (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00008712
CAS Number: 140-11-4
EINECS: 205-399-7
Merck Index: 13,01125
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27ea0ed6d6c01a29d47929aa1924777fb6441870&vid=2204a088bca1773d2d956393c1840ab1da347741&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR