Abietic acid ≥80.0% (by GC)

Supplier: TCI America

A0001-25G A0001-500G
TCA0001-025GEA 49.82 USD
TCA0001-025G TCA0001-500G
Abietic acid ≥80.0% (by GC)
Abietic acid

CAS Number: 514-10-3
MDL Number: MFCD03423567
Molecular Formula: C20H30O2
Molecular Weight: 302.46
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Melting point (°C): 164
Specific rotation [a]20/D: -75 deg (C=1, EtOH)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Diterpenes
Other Product Names / Synonyms: Rosin Acid, Sylvic Acid

Formula: C₂₀H₃₀O₂
Melting Pt: 164 °C
MDL Number: MFCD03423567
CAS Number: 514-10-3
EINECS: 208-178-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a00a2abc0e075590cc34e4e5022159c9ab927de&vid=6a00a2abc0e075590cc34e4e5022159c9ab927de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR