Methyl isobutyrylacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-methyl-3-oxopentanoate, Methyl 4-methyl-3-oxovalerate

L16216-14 L16216-22
AAAL16216-14EA 48.8 USD
AAAL16216-14 AAAL16216-22
Methyl isobutyrylacetate 98%
Methyl isobutyrylacetate

Purity: 98% CAS No.: 42558-54-3 Molecular Formula: C7H12O3 Formula Weight: 144.17 Flash Point: 79°(174°F) Refractive Index: 1.4265 Melting Point: -75° Boiling Point: 192° Beilstein Reference: 1857823 MDL No.: MFCD00040499

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 192 °C
Melting Pt: –75 °C
Density: 1.010
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040499
CAS Number: 42558-54-3
EINECS: 418-900-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR