Methyl oxalyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate

L13258-14 L13258-06 L13258-22
AAAL13258-14EA 135 USD
AAAL13258-14 AAAL13258-06 AAAL13258-22
Methyl oxalyl chloride 97%
Methyl oxalyl chloride

Purity: 97% CAS No.: 5781-53-3 Molecular Formula: C3H3ClO3 Formula Weight: 122.51 Flash Point: 46°(114°F) Refractive Index: 1.4190 Odor: Acrid Boiling Point: 115-118° Beilstein Reference: 1071541 UN No.: 2920 MDL No.: MFCD00000705

Formula: C₃H₃ClO₃
MW: 122.51 g/mol
Boiling Pt: 115…118 °C
Density: 1.332
Flash Pt: 46 °C(114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000705
CAS Number: 5781-53-3
EINECS: 227-307-4
UN: 2920
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR