Benzofurazan 97%

Supplier: Thermo Scientific Chemicals
Warning

L11229-03 L11229-06
AAAL11229-03EA 39 USD
AAAL11229-03 AAAL11229-06
Benzofurazan 97%
Benzofurazan

Purity: 97% CAS No.: 273-09-6 Molecular Formula: C6H4N2O Formula Weight: 120.11 Flash Point: 76°(169°F) Melting Point: 52-55° Boiling Point: 80°/20mm Beilstein Reference: 112261 MDL No.: MFCD00068064

Formula: C₆H₄N₂O
Boiling Pt: 80 °C/20 mm
Melting Pt: 52…55 °C
Flash Pt: 76 °C(169 °F)
Storage Temperature: Ambient
MDL Number: MFCD00068064
CAS Number: 273-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR