Isopropylmalonic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Isopropylmalonic acid
L10171-06
L10171-14
AAAL10171-06EA
75.7
USD
AAAL10171-06
AAAL10171-14
Isopropylmalonic acid 98+%
Isopropylmalonic acid
Purity: 98+%
CAS No.: 601-79-6
Molecular Formula: C6H10O4
Formula Weight: 146.14
Melting Point: 88-90°
Beilstein Reference: 1723234
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Melting Pt: 88…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021717 CAS Number: 601-79-6 EINECS: 210-008-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...