Methyl mercaptoacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl thioglycolate

L03783-36 L03783-22
AAAL03783-36EA 83.6 USD
AAAL03783-36 AAAL03783-22
Methyl mercaptoacetate 98%
Methyl mercaptoacetate

Purity: 98% CAS No.: 2365-48-2 Molecular Formula: C3H6SO2 Formula Weight: 106.14 Flash Point: 56°(132°F) Refractive Index: 1.4660 Odor: Stench Melting Point: -3° Boiling Point: 148-149° Beilstein Reference: 506259 UN No.: 1228 MDL No.: MFCD00004873

Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 148…149 °C
Melting Pt: –3 °C
Density: 1.163
Flash Pt: 56 °C (132 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004873
CAS Number: 2365-48-2
EINECS: 219-121-7
UN: 1228
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR