8-Bromoguanosine hydrate 97%

Supplier: Thermo Scientific Chemicals

L02992-03 L02992-06
AAAL02992-03EA 21 USD
AAAL02992-03 AAAL02992-06 AAAL02992-14
8-Bromoguanosine hydrate 97%
8-Bromoguanosine hydrate

Purity: 97% CAS No.: 332359-99-6 Molecular Formula: C10H12BrN5O5.xH2O Formula Weight: 362.14(anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 578725 Optical Rotation: -30° (c=2 in 1:1 DMSO/water) MDL No.: MFCD00150531

Formula: C₁₀H₁₂BrN₅O₅·H₂O
MW: 362.14 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150531
CAS Number: 332359-99-6
EINECS: 223-677-6

Order Now

Specification Test Results

Optical rotation -30° (c=2 in DMSO/water, 1:1)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR