Acridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L01657-06 L01657-14
AAAL01657-06EA 77.08 USD
AAAL01657-06 AAAL01657-14
Acridine 97%
Acridine

Purity: 98+% CAS No.: 260-94-6 Molecular Formula: C13H9N Formula Weight: 179.22 Melting Point: 107-111° Boiling Point: 346° Merck Reference: 14,122 UN No.: 2713 MDL No.: MFCD00005025

Formula: C₁₃H₉N
Boiling Pt: 346 °C
Melting Pt: 107…111 °C
Density: 1.005
Storage Temperature: Ambient
MDL Number: MFCD00005025
CAS Number: 260-94-6
EINECS: 205-971-6
UN: 2713
ADR: "6.1",III
Merck Index: 14,122

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR