2-Methyl-1,3-dithiane 98%
Supplier: Thermo Scientific Chemicals
Warning
|
L00209-06
L00209-14
AAAL00209-06EA
206
USD
AAAL00209-06
AAAL00209-03
AAAL00209-14
2-Methyl-1,3-dithiane 98%
2-Methyl-1,3-dithiane
Purity: 98%
CAS No.: 6007-26-7
Molecular Formula: C5H10S2
Formula Weight: 134.25
Flash Point: 82°(179°F)
Refractive Index: 1.5615
Boiling Point: 78-80°/9mm
Beilstein Reference: 102766
MDL No.: MFCD00014652
Formula:
C₅H₁₀S₂ MW: 134.27 g/mol Boiling Pt: 78…80 °C/9 mm Density: 1,212 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014652 CAS Number: 6007-26-7 EINECS: 227-859-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...