Benzo[b]thiophene-3-carboxylic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Benzothiophene-3-carboxylic acid

H50105-03 H50105-06 H50105-14
AAAH50105-03EA 88 USD
AAAH50105-03 AAAH50105-06 AAH50105-14
Benzo[b]thiophene-3-carboxylic acid 96%
Benzo[b]thiophene-3-carboxylic acid

Purity: 96% CAS No.: 5381-25-9 Molecular Formula: C9H6O2S Formula Weight: 178.22 Melting Point: 174-176° MDL No.: MFCD01846406

Formula: C₉H₆O₂S
MW: 178.22 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD01846406
CAS Number: 5381-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR