Dimethylamine 2 M in methanol

Supplier: Thermo Scientific Chemicals
Danger

H27261-AC H27261-AE
AAH27261-ACEA 53 USD
AAH27261-AC AAH27261-AE
Dimethylamine 2 M in methanol
Dimethylamine

CAS No.: 124-40-3 Molecular Formula: C2H7N Formula Weight: 45.08 Flash Point: 5°(41°F) Refractive Index: 1.3350 Beilstein Reference: 605257 Merck Reference: 14,3228 UN No.: 3286 MDL No.: MFCD00008288

Formula: C₂H₇N
MW: 45.08 g/mol
Density: 0.775
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 3286
ADR: 3,II
Merck Index: 14,3228

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR