1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
PYR14-TFSI, N-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulphonyl)imide
H27177-06
H27177-18
AAH27177-06EA
107
USD
AAH27177-06
AAH27177-18
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide 98%
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide
Purity: 98%
CAS No.: 223437-11-4
Molecular Formula: C11H20F6N2O4S2
Formula Weight: 422.42
MDL No.: MFCD07784447
Formula:
C₁₁H₂₀F₆N₂O₄S₂ MW: 422.41 Storage Temperature: Ambient |
MDL Number:
MFCD07784447 CAS Number: 223437-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...