(R)-(-)-4-Phenyl-2-oxazolidinone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-4-Phenyloxazolidin-2-one
H27171-06
H27171-03
AAH27171-06EA
138
USD
AAH27171-06
AAH27171-03
(R)-(-)-4-Phenyl-2-oxazolidinone 98%
(R)-(-)-4-Phenyl-2-oxazolidinone
Purity: 98%
CAS No.: 90319-52-1
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: 129-132°
Optical Rotation: -55° (c=2 in chloroform)
Formula:
C₉H₉NO₂ MW: 163.18 g/mol Melting Pt: 129…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192393 CAS Number: 90319-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...