1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, IPr.HCl

H27150-06 H27150-03
AAH27150-06EA 596 USD
AAH27150-06 AAH27150-03
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride 97%
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

Purity: 97% CAS No.: 250285-32-6 Molecular Formula: C27H37ClN2 Formula Weight: 425.05 Melting Point: ca 280° dec. MDL No.: MFCD02684545

Formula: C₂₇H₃₇ClN₂
MW: 425.05 g/mol
Melting Pt: ca 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02684545
CAS Number: 250285-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR