(±)-2-Phenylpiperazine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H26502-06
H26502-14
AAH26502-06EA
127
USD
AAH26502-06
AAH26502-14
(±)-2-Phenylpiperazine 96%
(±)-2-Phenylpiperazine
Purity: 96%
CAS No.: 5271-26-1
Molecular Formula: C10H14N2
Formula Weight: 162.24
Melting Point: 83-87°
UN No.: 3259
MDL No.: MFCD01871362
Formula:
C₁₀H₁₄N₂ MW: 162.23 g/mol Melting Pt: 83…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD01871362 CAS Number: 5271-26-1 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...