3-Chloro-5-fluoroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26338-03
H26338-06
AAH26338-03EA
156
USD
AAH26338-03
AAH26338-06
3-Chloro-5-fluoroaniline 97%
3-Chloro-5-fluoroaniline
Purity: 97%
CAS No.: 4863-91-6
Molecular Formula: C6H5ClFN
Formula Weight: 145.56
Refractive Index: 1.562-1.564
Odor: Characteristic
UN No.: 2810
MDL No.: MFCD03407962
Formula:
C₆H₅ClFN MW: 145.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03407962 CAS Number: 4863-91-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...