3-Chloro-5-fluorophenol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H26195-03
H26195-06
AAH26195-03EA
86.4
USD
AAH26195-03
AAH26195-06
3-Chloro-5-fluorophenol 98%
3-Chloro-5-fluorophenol
Purity: 98%
CAS No.: 202982-70-5
Molecular Formula: C6H4ClFO
Formula Weight: 146.55
Refractive Index: 1.537
Formula:
C₆H₄ClFO Storage Temperature: Ambient |
MDL Number:
MFCD00042212 CAS Number: 202982-70-5 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...