5-Chloro-2-fluorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26187-14
H26187-03
H26187-06
AAH26187-14EA
477
USD
AAH26187-14
AAH26187-03
AAH26187-06
5-Chloro-2-fluorobenzaldehyde 97%
5-Chloro-2-fluorobenzaldehyde
Purity: 97%
CAS No.: 96515-79-6
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Refractive Index: 1.5410
Melting Point: 29-32°
MDL No.: MFCD03094518
Formula:
C₇H₄ClFO Melting Pt: 29…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094518 CAS Number: 96515-79-6 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...