3-(3,4-Difluorophenyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(3,4-Difluorophenyl)propanoic acid

H26185-06 H26185-03
AAH26185-06EA 63.2 USD
AAH26185-06 AAH26185-03
3-(3,4-Difluorophenyl)propionic acid 98%
3-(3,4-Difluorophenyl)propionic acid

Purity: 98% CAS No.: 161712-75-0 Molecular Formula: C9H8F2O2 Formula Weight: 186.16 Melting Point: 49-53° MDL No.: MFCD00799520

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00799520
CAS Number: 161712-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR