2-Fluoroisobutyric acid 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H25837-03
H25837-MD
AAH25837-03EA
155
USD
AAH25837-03
AAH25837-MD
2-Fluoroisobutyric acid 95%
2-Fluoroisobutyric acid
Purity: 95%
CAS No.: 63812-15-7
Molecular Formula: C4H7FO2
Formula Weight: 106.10
Refractive Index: 1.3850
Boiling Point: 57-58°/10mm
UN No.: 2922
MDL No.: MFCD03452788
Formula:
C₄H₇FO₂ MW: 106.1 g/mol Boiling Pt: 57…58 °C/10 mm Storage Temperature: Ambient |
MDL Number:
MFCD03452788 CAS Number: 63812-15-7 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...