2,2-Difluorobutyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Difluorobutanoic acid
H25830-03
H25830-MD
AAH25830-03EA
136
USD
AAH25830-03
AAH25830-MD
2,2-Difluorobutyric acid 97%
2,2-Difluorobutyric acid
Purity: 97%
CAS No.: 2366-62-3
Molecular Formula: C4H6F2O2
Formula Weight: 124.09
Refractive Index: 1.3645
Melting Point: 7-9°
Boiling Point: 100-101°/8.5mm
UN No.: 3265
MDL No.: MFCD03093765
Formula:
C₄H₆F₂O₂ Boiling Pt: 100…101 °C/8.5 mm Melting Pt: 7…9 °C Density: 1.220 Storage Temperature: Ambient |
MDL Number:
MFCD03093765 CAS Number: 2366-62-3 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...