(S)-(+)-4-Penten-2-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(+)-1-Penten-4-ol, (S)-(+)-Allyl methyl carbinol, (S)-Pent-4-en-2-ol
B25728-03
B25728-06
AAB25728-03EA
161
USD
AAB25728-03
AAB25728-06
(S)-(+)-4-Penten-2-ol 97%
(S)-(+)-4-Penten-2-ol
Purity: 97%
CAS No.: 55563-79-6
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 30°(86°F)
Refractive Index: 1.4250
Boiling Point: 115-116°
Optical Rotation: +5.2° (c=1 in chloroform)
UN No.: 1987
MDL No.: MFCD03427200
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 115…116 °C Density: 0.837 Flash Pt: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03427200 CAS Number: 55563-79-6 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...