(R)-(-)-4-Penten-2-ol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(-)-1-Penten-4-ol, (R)-2-Hydroxypent-4-ene, (R)-(-)-Allyl methyl carbinol

B25659-03 B25659-06
AAB25659-03EA 173 USD
AAB25659-03 AAB25659-06
(R)-(-)-4-Penten-2-ol 95%
(R)-(-)-4-Penten-2-ol

Purity: 95% CAS No.: 64584-92-5 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 30°(86°F) Refractive Index: 1.4240 Boiling Point: 115-116° Optical Rotation: -5.2° (c=1 in chloroform) UN No.: 1987 MDL No.: MFCD03701536

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 115…116 °C
Density: 0.837
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD03701536
CAS Number: 64584-92-5
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR