2-Chloro-5-fluoroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25618-03
B25618-06
B25618-14
AAB25618-03EA
40.7
USD
AAB25618-03
AAB25618-06
AAB25618-14
2-Chloro-5-fluoroaniline 97%
2-Chloro-5-fluoroaniline
Purity: 97%
CAS No.: 452-83-5
Molecular Formula: C6H5ClFN
Formula Weight: 145.56
Refractive Index: 1.5560
Odor: Characteristic
Boiling Point: 211°
UN No.: 2811
MDL No.: MFCD00084852
Formula:
C₆H₅ClFN MW: 145.56 g/mol Boiling Pt: 211 °C Melting Pt: ca 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084852 CAS Number: 452-83-5 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...