5-Iodo-1,3-dimethyluracil 99%

Supplier: Thermo Scientific Chemicals
Warning

B25173-06 B25173-03
AAAB25173-06EA 211 USD
AAAB25173-06 AAAB25173-14 AAAB25173-03
5-Iodo-1,3-dimethyluracil 99%
5-Iodo-1,3-dimethyluracil

Purity: 99% CAS No.: 40738-83-8 Molecular Formula: C6H7IN2O2 Formula Weight: 266.04 Melting Point: 225° dec. MDL No.: MFCD00192228

Formula: C₆H₇IN₂O₂
MW: 266.04 g/mol
Melting Pt: 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00192228
CAS Number: 40738-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR