Bismut(III) gallate basic hydrate

Supplier: Thermo Scientific Chemicals

Synonyms: Bismuth subgallate hydrate, 2,7-Dihydroxy-1,3,2-benzodioxabismole-5-carboxylic acid hydrate, Dermatol hydrate

B24187-22 B24187-36
AAAB24187-22EA 39.6 USD
AAAB24187-22 AAAB24187-36
Bismut(III) gallate basic hydrate
Bismut(III) gallate basic hydrate

CAS No.: 99-26-3 Molecular Formula: C7H5BiO6.xH2O Formula Weight: 394.10(anhy) MDL No.: MFCD01632692

Formula: C₇H₅BiO₆·xH₂O
MW: 394.09 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD01632692
CAS Number: 342406-26-2
EINECS: 202-742-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR