Copper(II) acetate, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

B23615-14 B23615-22 B23615-36
AAB23615-14EA 124.84 USD
AAB23615-14 AAB23615-22 AAB23615-36
Copper(II) acetate, anhydrous 98%
Copper(II) acetate

Purity: 98% CAS No.: 142-71-2 Molecular Formula: C4H6CuO4 Formula Weight: 181.63 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Blue-green Odor: Odorless Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00008690

Formula: CuCH₃CO₂
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008690
CAS Number: 142-71-2
EINECS: 205-553-3
UN: 3260
ADR: 8,III
Merck Index: 14,2624

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR