O-p-Tolyl chlorothioformate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
O-4-Methylphenyl chlorothioformate, p-Tolyl chlorothionoformate
B23501-09
B23501-18
AAAB23501-09EA
80
USD
AAAB23501-09
AAAB23501-18
O-p-Tolyl chlorothioformate 97%
O-p-Tolyl chlorothioformate
Purity: 97%
CAS No.: 937-63-3
Molecular Formula: C8H7ClOS
Formula Weight: 186.66
Storage Temperature: 2°C to 8°C
Flash Point: 96°(204°F)
Refractive Index: 1.5740
Boiling Point: 70-72°/1mm
Beilstein Reference: 1448986
UN No.: 3265
MDL No.: MFCD00014466
Formula:
C₈H₇ClOS MW: 186.66 g/mol Boiling Pt: 70…72 °C (1 mmHg) Density: 1.210 Flash Pt: 96 °C (204 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014466 CAS Number: 937-63-3 EINECS: 213-333-3 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...