Potassium tetrachloroaurate hydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gold potassium chloride hydrate, Potassium tetrachloroaurate(III) hydrate, Potassium gold(III) chloride hydrate

B23292-03 B23292-06
AAB23292-03EA 149 USD
AAB23292-03 AAB23292-06
Potassium tetrachloroaurate hydrate 99%
Potassium tetrachloroaurate hydrate

Purity: 99% CAS No.: 27988-75-6 Molecular Formula: AuCl4K.xH2O Formula Weight: 377.88(anhy) Melting Point: 357° dec. Merck Reference: 14,7681 MDL No.: MFCD00043088

Formula: AuCl₄K·xH₂O
MW: 377.88 g/mol (anhy)
Melting Pt: 357 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043088
CAS Number: 27988-75-6
EINECS: 237-190-1
UN: 3260
ADR: 8,III
Merck Index: 14,7681

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR