(±)-Tetrahydrofurfurylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Tetrahydrofurfurylamine

B23068-36 B23068-22
AAB23068-36EA 134 USD
AAB23068-36 AAB23068-22
(±)-Tetrahydrofurfurylamine 97%
(±)-Tetrahydrofurfurylamine

Purity: 97% CAS No.: 4795-29-3 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 45°(113°F) Refractive Index: 1.4550 Boiling Point: 153-154° Beilstein Reference: 102729 UN No.: 2943 MDL No.: MFCD00005373

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 153…154 °C
Density: 0.98
Flash Pt: 45 °C(113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005373
CAS Number: 4795-29-3
EINECS: 225-351-9
UN: 2943
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR