Zinc D-gluconate hydrate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Gluconic acid Zzinc(II) salt hydrate, Zinc(II) gluconate hydrate, Zinc gluconate hydrate

B23022-22 B23022-36
AAB23022-22EA 56 USD
AAB23022-22 AAB23022-36
Zinc D-gluconate hydrate 97%
Zinc D-gluconate hydrate

Purity: 97% CAS No.: 4468-02-4 Molecular Formula: C12H22O14Zn.xH2O Formula Weight: 455.69(anhy) Odor: Odorless pH: 6.5 Merck Reference: 14,4456 MDL No.: MFCD00868110

Formula: C₁₂H₂₂O₁₄Zn·xH₂O
MW: 455.69 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00868110
CAS Number: 4468-02-4
EINECS: 224-736-9
Merck Index: 14,4456

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR