Strontium bromide hexahydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

B22553-22 B22553-36
AAB22553-22EA 70.85 USD
AAB22553-22 AAB22553-36
Strontium bromide hexahydrate 99%
Strontium bromide hexahydrate

m.p. 88° (rapidly heated) , 180° -6H2O


Purity: 99% CAS No.: 7789-53-9 Molecular Formula: Br2Sr.6H2O Formula Weight: 355.53 (247.44anhy) Odor: Odorless Merck Reference: 14,8837 MDL No.: MFCD00150361

Formula: Br₂Sr·6H₂O
Density: 2.386
Storage Temperature: Ambient
MDL Number: MFCD00150361
CAS Number: 7789-53-9
EINECS: 233-969-5
Merck Index: 14,8837

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR