4,4,4-Trifluorobutyric acid 97%

Supplier: Thermo Scientific Chemicals
Danger

B22186-03 B22186-06
AAB22186-03EA 54.2 USD
AAB22186-03 AAB22186-06
4,4,4-Trifluorobutyric acid 97%
4,4,4-Trifluorobutyric acid

Purity: 97% CAS No.: 406-93-9 Molecular Formula: CF3CH2CH2CO2H Formula Weight: 142.08 Melting Point: 29-32° Boiling Point: 166-167° UN No.: 3261 MDL No.: MFCD00077604

Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
Boiling Pt: 166…167 °C
Melting Pt: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00077604
CAS Number: 406-93-9
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR