Calix[8]arene 97%

Supplier: Thermo Scientific Chemicals
Warning

B21666-03 B21666-06
AAAB21666-03EA 94.7 USD
AAAB21666-03 AAAB21666-06
Calix[8]arene 97%
Calix[8]arene

Purity: 97% CAS No.: 82452-93-5 Molecular Formula: C56H48O8 Formula Weight: 848.99 Melting Point: 450° MDL No.: MFCD00143914

Formula: C₅₆H₄₈O₈
MW: 848.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143914
CAS Number: 82452-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR