Indomethacin 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19910-06
A19910-14
A19910-22
AAA19910-06EA
63.2
USD
AAA19910-06
AAA19910-14
AAA19910-22
Indomethacin 98%
Indomethacin
Purity: 98%
CAS No.: 53-86-1
Molecular Formula: C19H16ClNO4
Formula Weight: 357.8
Melting Point: 158-161°
Beilstein Reference: 497341
Merck Reference: 14,4968
UN No.: 2811
MDL No.: MFCD00057095
Formula:
C₁₉H₁₆ClNO₄ MW: 357.80 g/mol Melting Pt: 158…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00057095 CAS Number: 53-86-1 EINECS: 200-186-5 UN: 2811 ADR: 6.1,II Merck Index: 14,4968 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...