N,N-Dimethylglycine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid
A19670-14
A19670-22
A19670-06
AAA19670-14EA
285
USD
AAA19670-14
AAA19670-22
AAA19670-06
N,N-Dimethylglycine 98+%
N,N-Dimethylglycine
Purity: 98+%
CAS No.: 1118-68-9
Molecular Formula: C4H9NO2
Formula Weight: 103.12
Odor: Odorless
pH: 6-7
Melting Point: 179-183°
Merck Reference: 14,3244
MDL No.: MFCD00004283
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Melting Pt: 179…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004283 CAS Number: 1118-68-9 EINECS: 214-267-8 Merck Index: 14,3244 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...