L(+)-α-Phenylglycine 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phg-OH, L-(+)-2-Phenylglycine

A19360-36 A19360-14 A19360-22
AAAA19360-36EA 299 USD
AAAA19360-36 AAA19360-14 AAA19360-22
L(+)-α-Phenylglycine 98+%
L(+)-α-Phenylglycine

Fieser: 17,278


Purity: 98+% CAS No.: 2935-35-5 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 288° dec. Beilstein Reference: 2208675 Merck Reference: 14,7291 Optical Rotation: +156° (c=1 in 1N HCl) MDL No.: MFCD00064403

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 288 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064403
CAS Number: 2935-35-5
EINECS: 220-909-8
Merck Index: 14,7291

Order Now

Specification Test Results

Optical rotation +156° (c=1 in 1N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR