(±)-α-Bromo-γ-butyrolactone 97%

Supplier: Thermo Scientific Chemicals
Warning

A17344-14 A17344-22
AAAA17344-14EA 47.4 USD
AAAA17344-14 AAAA17344-22
(±)-α-Bromo-γ-butyrolactone 97%
(±)-α-Bromo-γ-butyrolactone

Purity: 97% CAS No.: 5061-21-2 Molecular Formula: C4H5BrO2 Formula Weight: 164.99 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5080 Boiling Point: 137-139°/6mm Beilstein Reference: 107800 MDL No.: MFCD00005387

Formula: C₄H₅BrO₂
MW: 164.99 g/mol
Boiling Pt: 137 - 139 °C (8 hPa)
Density: 1.994 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005387
CAS Number: 5061-21-2
EINECS: 225-764-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR