Aurintricarboxylic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15905-06
A15905-14
A15905-22
AAA15905-06EA
51.25
USD
AAA15905-06
AAA15905-14
AAA15905-22
Aurintricarboxylic acid
Aurintricarboxylic acid
Inhibits protein-nucleic acid interactions
CAS No.: 4431-00-9
Molecular Formula: C22H14O9
Formula Weight: 422.35
Melting Point:
Formula:
C₂₂H₁₄O₉ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011663 CAS Number: 4431-00-9 EINECS: 224-628-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...