3-Methylbenzo[b]thiophene 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Methylbenzothiophene
A15772-06
A15772-14
A15772-03
AAA15772-06EA
165
USD
AAA15772-06
AAAA15772-14
AAA15772-03
3-Methylbenzo[b]thiophene 98%
3-Methylbenzo[b]thiophene
Purity: 98%
CAS No.: 1455-18-1
Molecular Formula: C9H8S
Formula Weight: 148.23
Flash Point: 104°(219°F)
Refractive Index: 1.6240
Melting Point: -40°
Boiling Point: 72-74°/2mm
Beilstein Reference: 111282
MDL No.: MFCD00040243
Formula:
C₉H₈S MW: 148.23 g/mol Boiling Pt: 72…74 °C/2 mm Melting Pt: –40 °C Density: 1.13 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040243 CAS Number: 1455-18-1 EINECS: 215-934-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...