β-Cyclodextrine hydrate

Supplier: Thermo Scientific Chemicals

A14529-22 A14529-06 A14529-14
AAA14529-22EA 189 USD
AAA14529-22 AAA14529-06 AAA14529-14
β-Cyclodextrine hydrate
β-Cyclodextrine hydrate

Fieser: 6,151 8,133 9,129 12,150 15,107 18,117 21,151


CAS No.: 68168-23-0 Molecular Formula: C42H70O35.xH2O Formula Weight: 1134.98(anhy) Melting Point: 288° dec. Beilstein Reference: 5915513 Merck Reference: 14,2718 Optical Rotation: +142° (c=1.5 in water) MDL No.: MFCD00150811

Formula: C₄₂H₇₀O₃₅·H₂O
MW: 1134.98 g/mol (anhy)
Melting Pt: 288 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150811
CAS Number: 68168-23-0
EINECS: 231-493-2
Merck Index: 14,2718

Order Now

Specification Test Results

Optical rotation +142° (c=1.5 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR