Ethyl carbazate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl hydrazinecarboxylate

A13860-30 A13860-0B A13860-18
AAA13860-30EA 185 USD
AAA13860-30 AAA13860-0B AAA13860-18
Ethyl carbazate 97%
Ethyl carbazate

Fieser: 1,360


Purity: 97% CAS No.: 4114-31-2 Molecular Formula: C3H8N2O2 Formula Weight: 104.11 Flash Point: 86°(186°F) Melting Point: 44-47° Boiling Point: 85-86°/7mm Beilstein Reference: 878265 UN No.: 2811 MDL No.: MFCD00007595

Formula: C₃H₈N₂O₂
MW: 104.11 g/mol
Boiling Pt: 85…86 °C/7 mm
Melting Pt: 44…47 °C
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007595
CAS Number: 4114-31-2
EINECS: 223-903-3
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR