Boc-Pro-OH 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
A13744-06
A13744-14
AAA13744-06EA
31.6
USD
AAA13744-06
AAA13744-14
Boc-Pro-OH 99%
Boc-Pro-OH
Purity: 99%
CAS No.: 15761-39-4
Molecular Formula: C10H17NO4
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Melting Point: 132-136°
Beilstein Reference: 15828
Optical Rotation: -61° (c=1 in acetic acid)
MDL No.: MFCD00037324
Formula:
C₁₀H₁₇NO₄ Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037324 CAS Number: 15761-39-4 EINECS: 239-848-3 |
Specification Test Results
Optical rotation | -61° (c=1 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...