(±)-2-Methylglutaric acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpentanedioic acid
A11823-14
A11823-06
A11823-22
AAA11823-14EA
234.45
USD
AAA11823-14
AAA11823-06
AAA11823-22
(±)-2-Methylglutaric acid 98%
(±)-2-Methylglutaric acid
Purity: 98%
CAS No.: 617-62-9
Molecular Formula: C6H10O4
Formula Weight: 146.14
Melting Point: 74-78°
Boiling Point: 214-215°/22mm
MDL No.: MFCD00002661
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Boiling Pt: 214…215 °C/22 mm Melting Pt: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002661 CAS Number: 617-62-9 EINECS: 210-521-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...