4'-Ethoxyacetanilide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
A11200-36
A11200-22
AAA11200-36EA
90.2
USD
AAA11200-36
AAA11200-22
4'-Ethoxyacetanilide 97%
4'-Ethoxyacetanilide
Purity: 97%
CAS No.: 62-44-2
Molecular Formula: C10H13NO2
Formula Weight: 179.22
Melting Point: 134-136°
Beilstein Reference: 1869238
Merck Reference: 14,7204
MDL No.: MFCD00009094
Formula:
C₁₀H₁₃NO₂ Melting Pt: 134…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009094 CAS Number: 62-44-2 EINECS: 200-533-0 Merck Index: 14,7204 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...