Diethyl-3-oxoglutarate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate

A10933-30 A10933-18
AAA10933-30EA 173 USD
AAA10933-30 AAA10933-18
Diethyl-3-oxoglutarate 96%
Diethyl-3-oxoglutarate

Purity: 96% CAS No.: 105-50-0 Molecular Formula: C9H14O5 Formula Weight: 202.21 Storage Temperature: 2°C to 8°C Flash Point: 86°(186°F) Refractive Index: 1.4400 Boiling Point: 134-135°/10mm Beilstein Reference: 640146 MDL No.: MFCD00009200

Formula: C₉H₁₄O₅
MW: 202.21 g/mol
Boiling Pt: 134…135 °C/10 mm
Density: 1.113
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009200
CAS Number: 105-50-0
EINECS: 203-302-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR