Potassium ethyl malonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Monoethyl malonate potassium salt, Ethyl potassium malonate

A10720-14 A10720-22 A10720-36
AAAA10720-14EA 68.89 USD
AAAA10720-14 AAAA10720-22 AAAA10720-36
Potassium ethyl malonate 98%
Potassium ethyl malonate

Fieser: 10,182

Formula: C₅H₇KO₄
MW: 170.21 g/mol
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035603
CAS Number: 6148-64-7
EINECS: 228-156-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR