Decaborane 98%

Supplier: Thermo Scientific Chemicals
Danger

087892-03 087892-06 087892-14
AA87892-03EA 82 USD
AA87892-03 AA87892-06 AA87892-14
Decaborane 98%
Decaborane

Slightly soluble in cold water. Hydrolyzes in hot water. Soluble in ethyl acetate, 1-Bromopropane, ethyl silicate, benzene, alcohol, and carbon tetrachloride


In rocket propellants, and as a catalyst in olefin polymerization


In rocket propellants, and as a catalyst in olefin polymerization

Formula: B₁₀H₁₄
Boiling Pt: 213 °C
Melting Pt: 99…103 °C
Density: 0.940
Storage Temperature: Ambient
MDL Number: MFCD00011314
CAS Number: 17702-41-9
EINECS: 241-711-8
UN: 1868
ADR: 4.1,II
Merck Index: 14,2845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR